missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep F(ab')2 Fragment To Mouse IgG, Biotin, Polyclonal Secondary Antibody, MP Biomedicals
Marca: MP Biomedicals 0855600
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
Host Species: Sheep
Species Reactivity: Ms
Enzyme Immunoassay, Immunohistochemistry, Western Blotting, Electron Microscopy
Specifica
IgG | |
Biotin | |
Affinity Purified | |
RUO | |
Mouse |
Polyclonal | |
Sheep | |
1 mL | |
Secondary |