Learn More
Euro Protection™ Shock-Proof Cap
Description
- Cap with a plastic shell for light impact protection
- Does not protect against falling objects (see protective industrial helmets complying with the EN397 standard)
- Outer fabric:100% cotton
- Shell in ABS
- Outer cushions: EVA
- Inner cushions: polyethylene
- Airholes: brass
- Duration: 3 years
Specifications
Specifications
| Certifications/Compliance | EN 812 |
| Construction | Plastic shell with EVA foam |
| Color | Black |
| For Use With (Application) | Light impact protection |
| Includes | Brass Airholes |
| Material | Cotton, ABS, EVA, Polyethylene, Brass |
| Product Type | Shock-proof Cap |