Learn More
Bohlender™ SICCO™ Extractor Hood
Product Code. 17117744
Change view
Click to view available options
Width (Metric):
900 mm
1200 mm
Width (Metric) Interior:
860 mm
1160 mm
Weight (Metric):
20 kg
23 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Width (Metric) Interior Weight (Metric) Unit Size
17117744 1200 mm 1160 mm 23 kg Each
17107744 900 mm 860 mm 20 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17117744 Supplier Bohlender™ Supplier No. V 196004
  • ONLINE EXCLUSIVE PRICE
    3969.79€
    1983.19€ / pezzi 1
    Risparmia 1986.60€
    50% Sconto
 Restricted Item
Spedizione stimata: 11-03-2027
Extractor Hood Panels made of polycarbonate with a top and integrated connector DN 160 made of polypropylene.
  • Aluminum frame with panels made of polycarbonate
  • Top panel with integrated connector for exhaust system made of polypropylene
  • Robust and light-weight aluminum construction
  • Transparent and shatter-proof panels made of 5 mm thick polycarbonate
  • Standard connection DN 160 for aspirator tube
  • Superb all-round visibility
  • Large front-opening for comfortable working
  • Top side made of polypropylene for a good chemical resistance
Specifications
Material Aluminum, Polycarbonate, Polypropylene
Depth (English) 24 in.
Depth (English) Interior 22 in.
Depth (Metric) 600 mm
Depth (Metric) Interior 560 mm
Description DN 160 Threaded Necks
Height (English) 35 in.
Height (English) Interior 34.6 in.
Height (Metric) 900 mm
Height (Metric) Interior 880 mm
Weight (English) 51 lb.
Weight (Metric) 23 kg
Width (English) 47 in.
Width (English) Interior 46 in.
Width (Metric) 1200 mm
Width (Metric) Interior 1160 mm