Learn More
Fisherbrand™ S 50 R Sieve Holder
Supports analyzing sieves for the S 50 R Ultrasonic Cleaning Unit. Fisherbrand™ S 50 R Sieve Holders facilitate the special “sieve cleaning” program using two ultrasonic modes alternately, creating powerful cleaning pulses and perfect results.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Made of stainless steel
Specifica
Ultrasonic Bath, S 50 R | |
Stainless Steel |