missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifica
Specifica
| Antigene | Siglec-2/CD22 |
| Applicazioni | Flow Cytometry |
| Classificazione | Monoclonal |
| Clone | MYG13 |
| Coniugato | HRP |
| Formulazione | PBS |
| Alias gene | B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 |
| Specie ospite | Mouse |
| Immunogeno | Human Raji cells |
| Metodo di purificazione | Protein A or G purified |
| Vedi altri risultati |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?