Description
- Error limits according to class B, DIN EN ISO 385
- Interchangeable keys
- Calibrated to deliver (TD, Ex)
- With glass-key in the titration and intermediate stopcock
- Automatic zeroing
- Intermediate stopcock to recirculate residual liquid
Specifications
Specifications
| Capacità (sistema metrico) | 25 mL |
| Graduazioni | 0.1 mL |
| Include | Without bottle |
| Valvola di chiusura | i-stopcock, amber lateral stopcock, PTFE |
| Warranty | 12 months |