Description
- Ultra-wraparound frame
- PC/TPR co-injected temples for greater comfort
- Panoramic field of vision
- FLEX 160° temples
Specifications
Specifications
| Certificazione/Conformità | CE, EN166 FT |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| N. per confezione | 1 ciasc. |
| Peso (sistema metrico) | 30 g |
| Lens Material | Policarbonato |