Learn More
Sartorius™ Silicone O-ring
Product Code. 15992610 Shop All Sartorius Products
Change view
Click to view available options
Size:
25 x 3mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15992610 25 x 3mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15992610 Supplier Sartorius™ Supplier No. 6980117
  • ONLINE EXCLUSIVE PRICE
    9.00€
    8.23€ / pezzi 1
    Risparmia 0.77€
    8% Sconto
In stock
Specifications
For Use With (Application) For use with 25mm Glass vacuum filter holder
Quantity 1Each
Product Type O-Ring
Material Silicone
Size 25 x 3mm