Learn More
Uvex Silv-Air Classic 2200 FFP2
Sfoglia Tutto Uvex Prodotti
Click to view available options
:
Silv-Air Classic
Descrizione
- Classic folding and cup-shaped respirators
- Fit all kinds of facial shape
- Headband loops and integrated sealing lips in all protection classes
- Comfortable to wear for a longer period of time without becoming bothersome.
Specifica
Specifica
| Tipo di aerosol | N - Not Resistant to Oil |
| Colore | White |
| Design Type | Cup/Molded |
| Monouso | Single-use |
| Tipo di filtro | FFP2 |
| Materiale | Polypropylene, Polyester |
| Product Type | General Purpose Particulate Respirator |
| Size | Medium |
| With Exhalation Valve | No |
| Senza lattice | Yes |
| Vedi altri risultati |