Descrizione
- Classic, white flat-fold respirators
- Fit all kinds of facial shape
- Headband loops and integrated sealing lips in all protection classes
- Comfortable to wear for a longer period of time without becoming bothersome
Specifica
Specifica
| Tipo di aerosol | N - Not Resistant to Oil |
| Colore | White |
| Design Type | Flat-fold |
| Monouso | Limited-use |
| Tipo di filtro | FFP3 |
| Materiale | Polypropylene, Polyester |
| Product Type | General Purpose Particulate Respirator |
| Size | Small |
| With Exhalation Valve | Sì |
| Senza lattice | No |
| Vedi altri risultati |