missing translation for 'onlineSavingsMsg'
Learn More

Silver rod, 3.175mm (0.125in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058559
Click to view available options
Quantità:
5 cm
30 cm
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Silver rod
Nota nome 3.175mm (0.125 in.) dia., Hard
Note percentuale saggio (metals basis)
Quantità 5 cm
Percent Purity 99.9%
Odore Characteristic
Peso ≈0.83g/cm
Forma Rod

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'