missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific CytoSep™ Funnel Chambers for Hettich Cyto-Systems
Use with Hettich Cyto-Systems to optimize lab throughput with multiple funnel options.
dietro preventivo
Specifica
Da utilizzare con (apparecchiatura) | Hettich Cyto-Systems |
---|---|
Tipo | One-funnel Chamber |
Descrizione
- Available in four sizes from 1-8mL
- Use the 1mL chamber: for small-volume samples of low cell contents
- The 2 and 4mL chambers are for cell-rich samples (pleura, ascites, bronchial washings)
- The 8mL chamber accommodates large-volume samples (urine)
- Filters for chambers available separately; see cat. no. 22-045-333
For CSF, Viral infections, Bronchial secretions
Specifica
Hettich Cyto-Systems | |
One-funnel Chamber |