Learn More
Simport™ Scientific Tamper Evident Screw Caps
For all applications requiring tamper evident seal for utmost security and sample integrity
Marca:  Simport™ Scientific T552-7WTP
Dettagli aggiuntivi : Peso : 0.75000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 16565211
  • 53.49€ / pezzi 1000
Spedizione stimata
20-04-2021
Aggiungi al carrello
Includes: Lip seal
Descrizione
Descrizione
  • Made of polyethylene
  • Features long skirt and super fast thread design
  • Can be removed or sealed with single turn
  • With leakproof seal
Specifica
Specifica
7 mL Serum-standing Non-sterile Transport Tubes
Polyethylene
Lip seal
Tamper Evident Screw Cap
White