Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Super Unleaded Gasoline 92 Octane | 8006-61-9 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifica
Specifica
| Materiale o nome chimico | Premium Unleaded Gasoline |
| Imballaggio | 2mL amber prescored glass ampule |
| CAS | 86290-81-5, 75-09-2 |
| CAS min. % | 0.1, 99.9 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS max % | 0.1, 99.9 |
| Da utilizzare con (apparecchiatura) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | MeCl |
| Quantità | 1 mL |
Product intended for laboratory use only