Learn More
Poulten Graf™ Siringa Luer-Lock monouso a due parti
Especially designed for use in human medicine. Poulten Graf™ Single Use Luer-Lock Two-Part Syringes offer quality at a good cost-performance ratio.
Marca: Poulten Graf™ 7 41510
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Produced in accordance with ISO 7886, ISO 7864 and ISO 8537
- PVC-free and non-toxic
- Sterilixed with ethylene oxide according to DIN 13098 and dispatched in blister packs
- Extremely tight with a transparent, smooth operating cylinder
- Screen printed graduation
Specifica
Sì | |
Polypropylene | |
Siringa Luer Lock a 2 parti | |
Confezione da 50 | |
Luer Lock | |
Sì |
CE | |
Graduazione serigrafata | |
Sì | |
30 mL | |
Sì |