Learn More
Kartell Slide Storing System
Description
- Each single drawer can hold up to 13 supports for slides (code 921) and transparent cover plates (code 920), to be ordered separately
- Each slides support can contain 100 microscope slides, easily extractable
- Each cavity is numbered
- The transparent lids guarantee an easy check of the contents, protecting the slides from dust and other foreign bodies
- The complete Storing System can therefore contain up to 5,200 slide
Specifica
Specifica
| Type | Slide Storage Cabinet |
| Slide Capacity | 5000 |
| For Use With (Equipment) | Classification and storage of microscope slides |
| Includes | 4 Fixed Feet |