Learn More
Micronova™ SlideMop™ Mop Heads, Irradiated
BIOPHARMA
Product Code. 16666172 Shop All Micronova Products
Change view
Click to view available options
Material:
Stainless Steel, Polyester
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
16666172 Stainless Steel, Polyester Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16666172 Supplier Micronova™ Supplier No. MZCR6316IR
  • ONLINE EXCLUSIVE PRICE
    216.00€
    206.40€ / pezzi 24
    Risparmia 9.60€
    4% Sconto
Spedizione stimata: 01-04-2027
Mop covers for use with the Micronova™ SlideMop™. The SlideMop is a simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover and lock into place with an effortless downward push.
  • Polyester mop covers for use with the the Micronova SlideMop™ Flat Head Mop. (For SlideMop frame, see QSM16.)
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersalManufactured for use in Controlled Environments, ISO 5 thru 9
  • Good chemical resistance
Specifications
Product Type Irradiated Mop Slipcover
Head Type Flat
Color White
Length (English) 16 in.
Sterility Sterile
Length (Metric) 41 cm
Autoclavable Not Autoclavable
Disposable Reusable
Care Instructions Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment) Stainless Steel Frame
For Use With (Application) Cleaning and Sanitization of Cleanroom Surfaces
Color-coded No
Material Stainless Steel, Polyester