Learn More
Thermo Scientific™ SLIPFREE Ferrules
Product Code. 10140643 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
PEEK
Vespel
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10140643 Vespel Each
10404783 PEEK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10140643 Supplier Thermo Scientific™ Supplier No. 36024
Spedizione stimata: 02-12-2026
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
  • Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
  • Kel-F and PEEK are offered for applications in which Vespel is not suitable
TRUSTED_SUSTAINABILITY
Specifications
Type Sealing Ferrule
Description Vespel sealing ferrule
For Use With (Equipment) SLIPFREE Connectors for HPLC Columns
Material Vespel