missing translation for 'onlineSavingsMsg'
Learn More

SLTM Antibody, Novus Biologicals™

Codice prodotto. 18268301 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18268301 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18268301 Fornitore Novus Biologicals N. del fornitore NBP147299

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

SLTM Polyclonal specifically detects SLTM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifica

Antigene SLTM
Applicazioni Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Concentrazione 0.2 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulazione TBS and 0.1% BSA with 0.09% Sodium Azide
Alias gene SAFB-like, transcription modulator
Simboli geni SLTM
Specie ospite Rabbit
Immunogeno The immunogen for this product maps to a region between residue 300 and 350 of human SAFB-like Transcription Modulator using the numbering given in entry NP_079031.2 (GeneID 79811).
Metodo di purificazione Affinity Purified
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 79811
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.