Learn More
Hamilton™ Stainless Steel Small Hub Removable Custom Needles
Codice prodotto. 12311583
Cambia vista
Click to view available options
Stile del punto dell'ago:
2, 3 or 4
2, 3, or 4
5 or AS
Dimensione della confezione:
pezzi 6
Calibro dell'ago:
20 G
25 G
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Stile del punto dell'ago unitSize Calibro dell'ago
12311583 2, 3 or 4 pezzi 6 -
10509822 2, 3 or 4 pezzi 6 -
11512525 2, 3 or 4 pezzi 6 -
12396289 5 or AS pezzi 6 -
12376439 2, 3, or 4 pezzi 6 25 G
12326449 2, 3, or 4 pezzi 6 20 G
12336299 5 or AS pezzi 6 20 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 12311583 Fornitore Hamilton™ N. del fornitore 780409
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Used for animal injections to limit tissue damage and sample loss.
  • Autoclavable
  • Provided in packs of 6
  • Can be used with removable (RN)-style syringes that are 100μL and smaller
  • Custom lengths and point styles are available
  • Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection
Specifica
Stile del punto dell'ago 2, 3 or 4