Learn More
Cole-Parmer™ Small Tough-Spot Labels
Product Code. 15127082 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Blue
Red
Unit Size:
1000 rolls
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15127082 Blue 1000 rolls
15182323 Red Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15127082 Supplier Cole-Parmer™ Supplier No. 6220704
Spedizione stimata: 01-02-2027
Withstand steam-autoclaving, boiling water baths, freezer temperatures, organic solvents, and caustic agents without peeling or falling off. Cole-Parmer™ Small Tough-Spot Labels are made of chemically inert polyester that strongly adheres to all plastics and other materials.
  • Pre-cut, peel off labels are the ideal size to fit microcentrifuge tubes and other laboratory containers.
  • Withstand temperatures as low as -20°C;-80°C when used with virgin polypropylene tubes.
  • Scuff- and smear-resistant.
Specifications
Color Blue
Material Polyester
Diameter (Metric) 0.375 mm
Autoclavable Autoclavable
Temperature Range (Metric) –80° when used with virgin polypropylene tubes. Otherwise, –20°C