Learn More
Thermo Scientific™ SMART Digest™ Pepsin Kits
Product Code. 16885951 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Bulk Resin (No Collection Plate)
with Collection Plate
with Filter and Collection Plate
with Magnetic Bead, Bulk Option (No Collection Plate)
with SOLAμ Collection Plate
Unit Size:
1 set
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
16885951 with Collection Plate 1 set
16815961 with Filter and Collection Plate 1 set
16825961 with SOLAμ Collection Plate 1 set
16895951 Bulk Resin (No Collection Plate) 1 set
16805961 with Magnetic Bead, Bulk Option (No Collection Plate) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16885951 Supplier Thermo Scientific™ Supplier No. 60109110
 Restricted Item
Spedizione stimata: 25-11-2026
Use these pepsin protein digestion kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results.
Use Thermo Scientific™ SMART Digest™ Pepsin Kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Pepsin Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
How SMART Digest Pepsin Kits work
Pepsin cleaves proteins preferentially at carboxylic groups of aromatic amino acids such as phenylalanine, tryptophan and tyrosine. It will not cleave at bonds containing valine, alanine or glycine. Pepsin is often used to generate antibody fragments. Pepsin is used to generate F(ab')2 fragments by removing only that portion of the Fc domain beyond the hinge region. The resulting F(ab')2 fragments are composed of two antibody-binding Fab' fragments connected by disulfides in the hinge region.
TRUSTED_SUSTAINABILITY
Specifications
Description with Collection Plate
Capture None
Cleavage Hydrophobic residues in P1 and P1' positions at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid
Enzyme Pepsin
Format Pre-filled PCR Tubes
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Digest Pepsin Kit
Includes SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate