Learn More
Thermo Scientific™ Smart-Vue™ Alert
Replaces the use of an analog USB model for issuing Smart-Vue wireless monitoring alerts via voice and/or SMS.
Specifica
| Tipo | Smart-Vue Alert |
|---|---|
| Tipo di display | No Display |
| Tipi di allarme | Voice, SMS |
| Software Type | SmartVue Client Version 2.2, SmartVue Server |
| Requisiti | 36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above. |
Descrizione
Smart-Vue™ Alert is an online alert platform, replacing the use of an analog USB modem for issuing alerts via voice and/or SMS. This is a requirement in many regions due to the phase-out of analog telephone lines. Smart-Vue Alert is a for-pay service with a yearly subscription model.
Feature summary- 12-month or 36-month options, time counting from the date of installation
- Module-based license options with same ranges as server licenses: 1-20, 21-50, 51-100, 101-unlimited
- Customer benefits from an unlimited number of alerts/calls for the allowed number of modules
- Smart-Vue Alert supports both SMS/Text and Voice alerts
- There is no “text-only” option (i.e. without voice alerts) at this time.
- Users cannot customize alert messages. This feature could be studied for a future release if necessary.
Specifica
| Smart-Vue Alert | |
| Voice, SMS | |
| 36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above. |
| No Display | |
| SmartVue Client Version 2.2, SmartVue Server | |
| Wireless Monitoring |