Learn More
Thermo Scientific™ Smart-Vue™ Client Server Software
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software and enjoy greater peace of mind.
Marca:  Thermo Scientific™ SV611-500-LSB
Dettagli aggiuntivi : Peso : 0.37000kg
Codice prodotto. 15330351
  • 4017.00€ / Pezzo
Spedizione stimata
10-07-2020
Aggiungi al carrello
Descrizione
Descrizione
Intelligent Plug & Play Simplicity
  • Installation Wizard for simplified, step-by-step process
  • Service Discovery Protocol (SDP) enables every module to send signal to receiver—identifies most efficient communication route based on RF signal strength
  • Module automatically establishes new path to receiver if no communication during 6-hour period
  • Back-up system with memory capacity up to 3000 points, with uninterruptible power for continuous data logging
Intuitive Dashboard
  • Daily usage is simplified
  • Single-click export of reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats
  • Flexible creation of graphs summarizing data from individual or multiple sensors
  • Visual depiction of RF signal strength an QoS as monitored by each module
Data Traceability & Security
  • Secure, customized levels of user access
  • Audit trails
  • Archived alarms
  • Password protection
  • Secure digital transmission and data back-ups
Numerous Configuration Capabilities
  • Combines RF and TCP/IP network technologies
  • Share equipment data over entire network instantly
  • Download simple plug-in to visualize equipment data readings from any workstation
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local sales representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifica
Specifica
Smart-Vue Software 2.0, 49-unlimited sensors
Software drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Software|Sensor
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Videos
Documenti
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.