Learn More
Motic™ Trinocular Head for SMZ-171 Stereo Microscope
Codice prodotto. 15126866
Cambia vista
Click to view available options
Potenza di ingrandimento:
7.5X to 50X
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Potenza di ingrandimento Dimensione della confezione
15126866 7.5X to 50X Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15126866 Supplier Motic™ Supplier No. 1100200600761
  • ONLINE EXCLUSIVE PRICE
    905.00€
    803.37€ / pezzi 1
    Risparmia 101.63€
    11% Sconto
View all promotions
Spedizione stimata: 13-01-2027
Regulatory: This product is under eco-contribution tax.
Combined with its expanded accessory line, Motic™ Trinocular Head for SMZ-171 Stereo Microscope is an ideal instrument for the demanding requirements for both routine and research fields.
  • For use with SMZ-171
  • Greenough zoom, 7.5X to 50X
  • 2 WF10x/23mm eyepices included
  • ESD feature (electrostatic discharge)
Specifications
Include Head only
Potenza di ingrandimento 7.5X to 50X
Tipi di obiettivo Achromatic
Field of View 23 mm
Da utilizzare con (apparecchiatura) SMZ171 StereoZoom Microscope
Intervallo di zoom Zoom 1:6,7
Warranty 5 years mechanical & optical
Product Type Trinocular Head for SMZ171
Modello SMZ-171-TH
Head Style Trinocular
Eyepiece WF 10X/23 mm
Quantità 1 Each