missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRK Polyclonal Antibody
GREENER_CHOICE

Product Code. 15787741 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantità:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
15787741 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15787741 Supplier Invitrogen™ Supplier No. PA559897

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VPQDLEDDLT ATPLSHATVP QSPARAADSV LNGHRSKGLC DSAKKDDLPE LAGPALSTVP PASLKPTASG RKCLFRVEED EEEDEEDKKP MSLSTQVVLR RKPSVTNRLT SRKSAPVLNQ IFEEGESDDE FDMDENLPPK LSRLKM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigene SNRK
Applicazioni Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 0.10 mg/mL
Coniugato Unconjugated
Formulazione PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRK
N. accesso geni Q63553, Q8VDU5, Q9NRH2
Alias gene 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312
Simboli geni SNRK
Specie ospite Rabbit
Immunogeno Recombinant protein corresponding to Human SNRK. Recombinant protein control fragment (Product #RP-89801).
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 170837, 20623, 54861
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.