Learn More
SNX24 Monoclonal Antibody (2A11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524452
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Peptide Sequence: MEVYIPSFRY EESDLERGYT VFKIEVLMNG RKHFVEKRYS EFHALHKKLK KCIKTPEIPS KHVRNWVPKV LEQRRQGLET YLQAVILENE ELPKLFLDFL NVRHLPSLPK AESCGSFDET ESEESSKLSH QPVLLFLGDP YVLPAASDFP NVVIEGVLHG IFYPHLQPR
SNX24 Monoclonal antibody specifically detects SNX24 in Human samples. It is validated for ELISA.Specifica
SNX24 | |
Monoclonal | |
Unconjugated | |
SNX24 | |
2810011K15Rik; 5730433I16Rik; AI451341; PRO1284; SBBI31; Snx24; sorting nexin 24; sorting nexin-24; sorting nexing 24; UNQ654/PRO1284 | |
Mouse | |
Protein A | |
RUO | |
28966 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
2A11 | |
PBS with no preservative; pH 7.4 | |
Q9Y343 | |
SNX24 | |
SNX24 (AAH10886, 1 a.a. ∽ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |