Learn More
Sodium Aminosalicylate Dihydrate, LoGiCal
Product Code. 30459895
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459895 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459895 Supplier LoGiCal Supplier No. LGCFOR1645.00
 Restricted Item
Spedizione stimata: 30-11-2026
Add to basket
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Especificaciones
Chemical Name or Material Sodium aminosalicylate dihydrate
CAS 6018-19-5
Analyte or Component Names Sodium aminosalicylate dihydrate
Shipping Condition Room Temperature
Molecular Formula C7 H6 N O3 . Na . 2 H2 O
Certifications/Compliance ISO 9001
Synonym Sodium aminosalicylate dihydrate, Sodium 4-amino-2-hydroxybenzoate dihydrate, Benzoic acid, 4-amino-2-hydroxy-, monosodium salt, dihydrate, Salicylic acid, 4-amino-, monosodium salt, dihydrate, Monosodium p-aminosalicylate dihydrate, Sodium 4-amino-2-hydroxybenzoate dihydrate, Sodium 4-aminosalicylate dihydrate, Sodium p-aminosalicylate dihydrate, p-Aminosalicylic acid sodium salt dihydrate, Benzoic acid, 4-amino-2-hydroxy-, sodium salt, hydrate (1:1:2), Mesalazine Impurity E as Sodium Salt Dihydrate
InChI Formula InChI=1S/C7H7NO3.Na.2H2O/c8-4-1-2-5(7(10)11)6(9)3-4;;;/h1-3,9H,8H2,(H,10,11);;2*1H2/q;+1;;/p-1
Solution Type Neat
SMILES O.O.[Na+].Nc1ccc(C(=O)[O-])c(O)c1
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
Industry Type Clinical Testing
Molecular Weight (g/mol) 211.15
Quantity 10 mg
Formula Weight 211.0457 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.