Learn More
Sodium Bitartrate Monohydrate, TRC
Specifications
Specifications
| Materiale o nome chimico | Sodium Bitartrate Monohydrate |
| CAS | 6131-98-2 |
| Formula molecolare | C4 H5 O6 . Na . H2 O |
| Quantità | 10 g |
| Sinonimo | Sodium Hydrogen Tartrate Monohydrate, (2R,3R)-2,3-Dihydroxybutanedioic Acid Sodium Salt Monohydrate, Sodium Tartaric Acid Monohydrate |
| InChI Formula | InChI=1S/C4H6O6.Na.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t1-,2-;;/m1../s1 |
| SMILES | O.[Na+].O[C@H]([C@@H](O)C(=O)[O-])C(=O)O |
| IUPAC Name | sodium;(2R,3R)-2,3,4-trihydroxy-4-oxobutanoate;hydrate |
| Molecular Weight (g/mol) | 190.084 |
| Peso formulazione | 190.009 |