Learn More
Sodium Cromoglicate, Tocris Bioscience™
Promotes ESCs/iPSCs differentiation into pancreatic endocrine islet cells
Marca: Tocris Bioscience 6424/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sodium Cromoglicate is a promotes transformation ESCs/iPSCs into pancreatic endocrine islet cells. Facilitates differentiation from endocrine precursors. Also inhibits mast cell degranulation and histamine release. Antiasthmatic.Specifica
| 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid] disodium salt | |
| 15826-37-6 | |
| >98% | |
| 50 mg |