missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Microprills, ≥98.5%, Honeywell™

Catalog Number p-7122041
Click to view available options
Quantità:
2.5 kg
6 x 2.5 kg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Microprills,≥98.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Sodium Hydroxide
Formula lineare NaOH
Numero UN 1823
Informazioni di solubilità Soluble in water, 1260g/L at 20°C
Quantità 2.5 kg
Peso formulazione 40g/mol
Percent Purity ≥98.5%
Imballaggio Poly bottle
Densità vapore >1 (vs air)
Pressione del vapore 3mmHg (37°C), 18mmHg (20°C)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'