Learn More
Sodium Hydroxide Solution, 30%, Chem Lab™
Product Code. 11923345
Change view
Click to view available options
Quantity:
5 L
Packaging:
5L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11923345 5 L 5L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11923345 Supplier Chem-Lab Supplier No. CL02.1407.5000
  • ONLINE EXCLUSIVE PRICE
    87.00€
    42.40€ / 5L
    Risparmia 44.60€
    51% Sconto
 Restricted Item
Spedizione stimata: 07-12-2026
Specifications
Color Colorless
Concentration 400g NaOH/l H2O
Density 1.33g/mL
Solubility Information Soluble in water
Quantity 5 L
Odor Odourless
Packaging 5L
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide 30 weight % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide