missing translation for 'onlineSavingsMsg'
Learn More

Sodium potassium (K2Na)-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™

Catalog Number p-7059576
Click to view available options
Quantità:
5 g
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
MFCD08705348
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
MFCD08705348

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Sodium potassium (K2Na)-silica gel, Stage I
Colore Black
Densità 0.7g/mL
Quantità 5 g
Numero UN UN2813
Sensibilità Moisture Sensitive
Concentration or Composition (by Analyte or Components) 35 to 40% alkali metal in silica gel
Forma fisica Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'