missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfite, ≥98% (RT), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 7757-83-7 |
| Linear Formula | Na2 SO3 |
| MDL Number | MFCD00003503 |
| Synonym | E221 |
| Solubility | Soluble in water |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 126.04 |
| Percent Purity | ≥98% (RT) |
| Grade | Analytical Standard |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?