missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99+%, Thermo Scientific Chemicals

Catalog Number p-7023102
Click to view available options
Quantità:
500 g
2500 g
10000 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1303-96-4
B4Na2O7·10H2O
MFCD00149193
Borax; Sodium borate (tetra) decahydrate
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1303-96-4
B4Na2O7·10H2O
MFCD00149193
Borax; Sodium borate (tetra) decahydrate

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

Solubility
Soluble in water, glycerol and ethanol. Insoluble in acids

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Sodium tetraborate decahydrate
Punto di fusione 75°C
pH 9.24
Quantità 500 g
Formula lineare Na2B4O7·10H2O
Indice di Merck 14,8590
Informazioni di solubilità Soluble in water,glycerol and ethanol. Insoluble in acids
Peso formulazione 381.37 (201.22 Anhydrous)
Percent Purity ≥99%
Odore Odorless
Densità 1.73
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • Sodium tetraborate decahydrate
Avvertenza
  • Pericolo
Categoria di pericolo
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Tossicità per la riproduzione Categoria 1B
Indicazioni di pericolo
  • H319-Provoca grave irritazione oculare.
  • H360FD-Può nuocere alla fertilità. Può nuocere al feto.
Consigli di prudenza
  • P201-Procurarsi istruzioni specifiche prima dell'uso.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P308+P313-IN CASO di esposizione o di possibile esposizione, consultare un medico.
Informazioni supplementari
  • MIXTURE LIST-Contiene: borax

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'