Learn More
Thermo Scientific™ Solid Door Option for Large Chamber
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Dimensione della confezione:
Pezzo
Descrizione
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
- Replaces standard outer glass door to protect samples against light
- Recommended for light sensitive applications
- Factory-installed option, please order in conjunction with environmental chamber
- Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
- Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifica
Specifica
| Descrizione | Solid door for large chamber, replaces standard outer glass door, factory installed |
| Da utilizzare con (apparecchiatura) | order with chamber models 3920, 3940, 3948, 3960, 3962 |
| Lighting | LED |
| Tipo | Solid Door Option |
| Door Style | Solid door for large chamber |
| Da utilizzare con (applicazione) | light sensitive applications |
| Include | Factory installed solid door |
Titolo del prodotto
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering