missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ A800 Grey TSR Glasses

Economical and comfortable, these safety glasses are affordably priced and suitable for use in a variety of application arenas. Honeywell Safety Products™ A800 Grey TSR Glasses are designed with a sporty style and a slim profile for workers with smaller faces.

Marca:  Honeywell Safety Products™ 1015367

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 11837080

  • 44.55€ / pezzi 10


Spedizione stimata
09-06-2021
Aggiungi al carrello

Descrizione

Descrizione

  • General use and protection against impact (45m/s).
  • Well suited for indoor or outdoor applications such as utilities, light and heavy manufacturing, bio tech, laboratories, clean rooms, ship building and automotive manufacturing.
  • Affordably priced and economical.
  • Slim profile for workers with smaller faces.
  • Sporty styling.
  • 9-base polycarbonate, abrasion resistant lenses.
  • 180° field of distortion-free vision.
  • Temples feature soft, padded inserts with extra flex.
  • Soft nosepiece conforms to various nose bridge shapes.
Specifica

Specifica

EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1790, EC Attestation Number EC1790
Gray
Anti-fog
Gray
Safety glasses
Administration. General use and protection against impact (45m/s). Well suited for indoor/outdoor applications such as utilities, light and heavy manufacturing, bio tech, laboratories, clean rooms, ship building and automotive manufacturing.
1 Ea.
Polycarbonate
Impact-resistant, Anti-fog
Videos
SDS
Documenti