Learn More
- Home
- Products
- Protein Analysis Reagents
- Dialysis and Desalting
- General Dialysis Supplies
- Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Product Code. 11571160| Product Code. | MWCO | Color Code | Length (Metric) | Unit Size |
|---|---|---|---|---|
| 11571160 | 100 kDa | Blue | 50 mm | Pack of 12 |
| 11531160 | 3.5 to 5 kDa | Black | 50 mm | Pack of 12 |
| 11541180 | 50 kDa | Violet | 160 mm | Pack of 12 |
| 11561170 | 100 kDa | Blue | 100 mm | Pack of 12 |
| 11581160 | 300 kDa | Amber | 50 mm | Pack of 12 |
| 11511160 | 0.1 to 0.5 kDa | Green | 50 mm | Pack of 12 |
| 11541160 | 8 to 10 kDa | Yellow | 50 mm | Pack of 12 |
| 11551160 | 20 kDa | Red | 50 mm | Pack of 12 |
| 11581170 | 0.1 to 0.5 kDa | Green | 160 mm | Pack of 12 |
| 11591170 | 1000 kDa | Pink | 100 mm | Pack of 12 |
| 11561160 | 50 kDa | Violet | 50 mm | Pack of 12 |
| 11495829 | 8 to 10 kDa | Yellow | 160 mm | Pack of 12 |
| 11571180 | 1000 kDa | Pink | 160 mm | Pack of 12 |
| 11501170 | 0.1 to 0.5 kDa | Green | 100 mm | Pack of 12 |
| 11501180 | 0.5 to 1 kDa | Orange | 160 mm | Pack of 12 |
| 11571170 | 300 kDa | Amber | 100 mm | Pack of 12 |
| 11551170 | 50 kDa | Violet | 100 mm | Pack of 12 |
| 11511170 | 0.5 to 1 kDa | Orange | 100 mm | Pack of 12 |
| 11591160 | 1000 kDa | Pink | 50 mm | Pack of 12 |
| 11561180 | 300 kDa | Amber | 160 mm | Pack of 12 |
| 11521160 | 0.5 to 1 kDa | Orange | 50 mm | Pack of 12 |
| 11541170 | 20 kDa | Red | 100 mm | Pack of 12 |
| 11521170 | 3.5 to 5 kDa | Black | 100 mm | Pack of 12 |
| 11531180 | 20 kDa | Red | 160 mm | Pack of 12 |
| 11837271 | 8 to 10 kDa | Yellow | 100 mm | Pack of 12 |
| 11827271 | 3.5 to 5 kDa | Black | 160 mm | Pack of 12 |
| 11551180 | 100 kDa | Blue | 160 mm | Pack of 12 |
-
380.00€
184.00€ / pezzi 12
Risparmia 196.00€
51% Sconto
Eseguire il login per visualizzare lo stock disponibile
Description
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
- Superior handling and leak protection
- Resealable screw cap and pipet for easy loading and total recovery
- Highest membrane purity, no heavy metals and sulfides
- Minimizes volume specific dilution
- 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
Specifications
| Product Type | Float-A-Lyzer G2 Dialysis Device |
| MWCO | 100 kDa |
| Membrane | Cellulose Ester |
| Diameter (English) Membrane | 0.39 in. |
| Diameter (English)Top | 0.9 in. |
| Diameter (Metric) Membrane | 10 mm |
| Diameter (Metric)Top | 23 mm |
| Length (English) | 1.9 in. |
| Length (Metric) | 50 mm |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |
| Color Code | Blue |
| Kit Contents | Disposable pipette for 5 and 10mL, flotation ring |
| Quantity | 12 Pack |
| For Use With (Application) | Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions |
| Show More Show Less |