Learn More
Molecular Devices SpectraMax™ iD5e Multi‐Mode uPlate AS with SoftMax Pro Software
Sfoglia Tutto Molecular Devices ProdottiDescription
- Designed for multi-mode detection workflows in microplate-based assays
- Supports a broad range of detection applications including complex assay workflows
- Enables flexible assay setup with tunable monochromator- and filter-based detection
- Supports biochemical and cell-based assay workflows across life science applications
- Provides enhanced detection capability for advanced and evolving assay requirements
- Includes preconfigured protocols to support common assay workflows
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Designed to support flexible and scalable laboratory workflows
Specifications
Specifications
| Temperatura incubatore | 4-Zone Incubation to 70°C |
| Velocità di misurazione | 11 sec. (96 wells), 22 sec. (384 wells) |
| Agitazione | Linear, Orbital, Double Orbital |
| Tipo di piastra | 6 to 384-well Plates |
| Tipo di rilevatore | PMT |
| Software Type | SoftMax Pro |
| Sorgente luminosa | Xenon Flash |
| Reading Options | Endpoint, Kinetic, Spectral Scanning |