Learn More
Molecular Devices SpectraMax™ L Microplate Reader with SoftMax Pro Software
Sfoglia Tutto Molecular Devices ProdottiDescrizione
- Designed for luminescence detection workflows in microplate-based assays
- Supports luminescence measurement applications for life science research
- Enables flexible assay setup with configurable detection parameters
- Supports biochemical and cell-based assay workflows across life science applications
- Provides programmable measurement workflows for routine and advanced assays
- Supports microplate-based screening applications across multiple formats
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Designed for sensitive detection of luminescent assay signals
Specifica
Specifica
| Tipo di rilevatore | Photomultiplier Tube (PMT) |
| Interfaccia | PC Control, Internal Software |
| Sorgente luminosa | LED |
| Tecnologia di misurazione | Luminescence (Top) |
| Tipo di piastra | 96 and 384-well Plates |
| Reading Options | Endpoint |
| Agitazione | None |
| Selezione della lunghezza d'onda | Filters (FL, Lum.) |
Titolo del prodotto
¿Detecta una oportunidad de mejora?