Learn More
Molecular Devices SpectraMax™ M3 Multimode Microplate Reader with SoftMax Pro Software
Sfoglia Tutto Molecular Devices ProdottiDescrizione
- Designed for multi-mode detection workflows in microplate-based assays
- Supports absorbance, fluorescence, and luminescence measurement applications
- Enables flexible assay setup with monochromator-based wavelength selection
- Supports biochemical and cell-based assay workflows across life science applications
- Supports endpoint, kinetic, and spectral measurement workflows
- Provides configurable detection settings for diverse assay requirements
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Designed for flexible and reliable operation in laboratory environments
Specifica
Specifica
| Temperatura incubatore | 4-Zone Incubation to 45°C |
| Velocità di misurazione | 11 sec. (96 wells), 22 sec. (384 wells) |
| Agitazione | Linear, Orbital |
| Tipo di piastra | 6 to 384-well Plates |
| Tipo di rilevatore | PMT |
| Software Type | SoftMax Pro |
| Sorgente luminosa | Xenon Flash |
| Reading Options | Endpoint, Kinetic, Spectral Scanning |