Learn More
Spectrum™ Spectra/Por™ Magnetic Weighted Closures
Product Code. 11550900
Change view
Click to view available options
Diameter (Metric):
23 mm
35 mm
55 mm
75 mm
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11550900 23 mm Pack of 2
11570900 55 mm Pack of 2
11560900 35 mm Pack of 2
11580900 75 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11550900 Supplier Spectrum™ Supplier No. 132760
  • ONLINE EXCLUSIVE PRICE
    103.00€
    91.18€ / pezzi 2
    Risparmia 11.82€
    11% Sconto
View all promotions
Spedizione stimata: 01-12-2026
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Magnetic Closure Type:
  • Made from polypropylene with a fluorocarbon coated magnetic SS bar
  • Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency
  • 4 sealing width sizes: 23mm, 35mm, 55mm and 75mm
  • Autoclavable
  • 10/pkg
  • Use with RC dialysis tubing only
  • Not for use with CE tubing
Specifications
Product Type Dialysis Tubing Closure
Membrane Regenerated Cellulose
Material Polypropylene
Diameter (Metric) 23 mm
Autoclavable Autoclavable
Closure Type Magnetic
Color Red
Temperature Range (English) 32°F to 250°F
Temperature Range (Metric) 0°C to 121°C
Kit Contents Magnetic fluorocarbon coated stainless steel bar
Quantity 2 Pack
For Use With (Application) Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency, use with RC dialysis tubing only, not for use with CE tubing