Learn More
3M™ Speedglas™ Inner Cover Plate
Product Code. 15819634 Shop All 3M Products
Change view
Click to view available options
Includes:
Marked 117 x 61
Marked 117 x 77
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15819634 Marked 117 x 77 Each
15809634 Marked 117 x 61 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15819634 Supplier 3M™ Supplier No. 7000000227
  • ONLINE EXCLUSIVE PRICE
    30.15€
    23.64€ / pezzi 1
    Risparmia 6.51€
    21% Sconto
Spedizione stimata: 05-02-2027
3M™ Speedglas™ Inner Cover Plate is designed to protect the inside surface of the filter from spark and spatter.
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
For Use With (Equipment) Inner Shield
Includes Marked 117 x 77
Product Type Cover Plate
Quantity 5/Pk.
Style 9100X