Learn More
3M™ Speedglas™ Outer Protection Plate 9100
Codice prodotto. 15809654 Sfoglia Tutto 3M Prodotti
Cambia vista
Click to view available options
Color: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
15809654 Color: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15809654 Fornitore 3M™ N. del fornitore 7100016691
  • ONLINE EXCLUSIVE PRICE
    47.40€
    36.78€ / pezzi 10
    Risparmia 10.62€
    22% Sconto
Spedizione stimata: 05-02-2027
3M™ Speedglas™ Outer Protection Plate 9100 is designed to protect the filter from spark and spatter.
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Specifica
Colore Clear
Product Type Helmet Outside Protection Plate