Learn More
Labconco™ SpillStopper™ Work Surface for 6 Ft. Protector™Premier™and Protector™XL Hoods
Codice prodotto. 17455902 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Include:
Right Rear
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Include Dimensione della confezione
17455902 Right Rear Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17455902 Fornitore Labconco™ N. del fornitore 9500601
  • ONLINE EXCLUSIVE PRICE
    2205.00€
    1744.08€ / pezzi 1
    Risparmia 460.92€
    20% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 12-01-2027
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow in the hood
  • Pre-drilled holes for venting and plumbing passthroughs provided
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 6 x 3 in. (15.2 x 7.6cm) cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
9500622
Not for use with Protector Solvent Storage Cabinets
Specifica
Da utilizzare con (apparecchiatura) Protector XL Laboratory Hood
Include Right Rear
Product Type Work Surface
Materiale Epoxy Resin
Da utilizzare con (applicazione) Laboratory Fume Hood Work