Learn More
Spiramycin, TRC
Product Code. 30136042
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136042 25 g 25g
30132707 5 g 5g
17940875 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136042 Supplier Toronto Research Chemicals Supplier No. TRCS68237325G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8025-81-8
Molecular Formula C43H74N2O14
Molecular Weight (g/mol) 843.05
Synonym Spiramycin, Antibiotic 799, Espiramicin, Foromacidin, IL 5902, NSC 55926, Provamycin, RP 5337, Rovamicina, Rovamycin, Rovamycine, Selectomycin, Sequamycin, Spiramycins, Stomamycin
IUPAC Name 2-((4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-(((2S,3R,4R,5S,6R)-5-(((2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-10-(((5S,6R)-5-(dimethylamino)-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxooxacyclohexadeca-11,13-dien-7-yl)acetaldehyde
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C43H74N2O14/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3/b13-12+,16-14+/t24?,25?,26?,27?,28?,29-,30-,31+,32-,34?,35-,36+,37+,38+,39-,40?,41-,42-,43?/m0/s1
Formula Weight 842.51
Percent Purity >95%
Chemical Name or Material Spiramycin
Grade HPLC