missing translation for 'onlineSavingsMsg'
Learn More

SPIRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17757-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897714

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in vesicle transport processes providing a novel link between actin organization and intracellular transport.
Specifica

Specifica

SPIRE2
Polyclonal
Unconjugated
SPIRE2
KIAA1832, Protein spire homolog 2, Spir 2, SPIR2, spire homolog 2 (Drosophila), SPIRE2
Rabbit
Antigen Affinity Chromatography
RUO
234857, 307925, 84501
-20°C
Liquid
Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K1S6, Q8WWL2
SPIRE2
SPIRE2 Fusion Protein Ag12117
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali