Learn More
Uvex Sport Occupational Black PU 65937 Low Shoe - Width 10
Offers first-class wearer comfort; low weight, climate-optimizes high-tech materials and ergonomic outsole
Marca:  Uvex 6593741
Dettagli aggiuntivi : Peso : 0.81200kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17242641
  • 125.00€ / 1 paio
Spedizione stimata: 08-11-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex 1 Sport Shoe 6593.7 O1 FO SRC
  • Modern, ultra-lightweight and flexible O1 occupational shoe without toe cap
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials are free of silicones plasticizers and other paint wetting impairment substances
  • Meets ESD specification with leakage resistance less than 35 megohms
  • Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
  • Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
  • Virtually seam-free construction made from high-tech materials to eliminate pressure points
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Soft, padding on collar and tongue
  • For light applications
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 6593.7
  • Standard EN ISO 20347:2012 O1 FO SRC
  • Sizes: 35 to 52
Specifica
Specifica
Black
41
Plain Toe
EN ISO 20347:2012
Narrow width
Yes
O1
Textile
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 1
No
SRC