Learn More
Gilac™ Square Bucket with Watertight Lid, 12 L
Codice prodotto. 17998511
Cambia vista
Click to view available options
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione della confezione
17998511 Pezzo
17908861 Pezzo
17998431 Pezzo
17978671 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 17998511 Fornitore Gilac™ N. del fornitore G615712
  • ONLINE EXCLUSIVE PRICE
    46.41€
    32.69€ / pezzi 1
    Risparmia 13.72€
    29% Sconto
Spedizione stimata: 28-10-2026
Square 12 L Bucket with Watertight Lid fit for food contact with watertight lid and pouring spout.
  • Bucket fit for food contact with watertight lid and pouring spout.
  • Airtight lid keeps flavours in and odours out.
  • Nesting when empty, no suction and stacking when lid is closed.
  • Enables liquid preparations to be stored and transported.
  • Graduation in litres.
  • Discharge area that prevents water retention when being cleaned and anti-wear skirt that lifts the bucket off the ground.
  • HDPE body and LDPE lid.
  • Stainless steel handle.
  • External dimensions include handle.
Specifica
Include Bucket, Handle, and Lid
Quantità 12 L