Learn More
Masterflex™ Stainless Steel Barbed Fitting NPT Male Pipe Adapter
Designed for quick and easy assembly without tools
Marca:  Masterflex™ 31210-33
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11841073
  • 65.34€ / Pezzo
Spedizione stimata
02-07-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Available in a variety of materials and configurations to meet your need
  • Adapts from a NPT male thread to a hose barb
Specifica
Specifica
1/2 NPT(M) x 5/8 in. Tubing ID
2 1/32 in.
5/8 in.
1/2 NPT(M)
1/2 NPT(M)
Stainless Steel
Barbed Fitting NPT Male Pipe Adapter
5/8 in.
5/8 in.
7/8 in.